Learn More
MilliporeSigma™ Calbiochem™ IκB Kinase Inhibitor Peptide, Cell-Permeable
Shop All MilliporeSigma ProductsDescription
- Packaged Under Inert Gas
- Peptide Sequence: Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Asp-Arg-His-Asp-Ser-Gly-Leu-Asp-Ser-Met-Lys-Asp-Glu-NH2
Specifications
Specifications
| Formulation | Supplied as a trifluoroacetate salt. |
| Description | 95% by HPLC |
| Product Type | Inhibitor |
| Target Proteins | LPS-induced IκB degradation by IKK in RAW 264.7 cells |
| Molecular Formula | C138H229N37O45S |
| Formula Weight | 3158.6g/mol |
| Solubility | PBS, pH 7.4 |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Lyophilized |
| Quantity | 1 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.