Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (±)-Ibuprofen

Catalog No. 4010031GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-100-31GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-100-31GM Supplier MilliporeSigma™ Supplier No. 4010031GM
Add to Cart
Add to Cart

A nonsteroidal anti-inflammatory drug (NSAID) that acts as a reversible and competitive inhibitor of cyclooxygenase 1 (COX-1) (IC50 = 4.85 μM).

A nonsteroidal anti-inflammatory drug (NSAID) that acts as a reversible and competitive inhibitor of cyclooxygenase 1 (COX-1) (IC50 = 4.85μM). Inhibits COX-2 at higher concentrations (IC50 = 223μM). Blocks aspirin inactivation of COX-1 (EC50 antagonism of 200μM aspirin = 290 nM). Shown to reduce the total Aβ secretion (Amyloid β40 and 42) in human neuronal cells and offers neuroprotection against glutamate-, nitric oxide-, and superoxide-induced damage. Reported to activate peroxisome proliferator-activated receptors (PPAR) α and γ in both CV-1 and C3H10T1/2 cells (∽100μM-500μM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by titration
Target Proteins COX-1
CAS 15687-27-1
Molecular Formula C13H18O2
Formula Weight 206.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.