Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ICMT Inhibitor II, FTPAT

Catalog No. 42035110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-035-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-035-110MG Supplier MilliporeSigma™ Supplier No. 42035110MG
Add to Cart
Add to Cart

The ICMT Inhibitor II, FTPAT controls the biological activity of ICMT.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Molecular Formula C26H31N3O2S
Formula Weight 449.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.