Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IDH2 Inhibitor, R140Q Somatic Mutant-Specific, AGI-6780

Catalog No. 5098800001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-988-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-988-00001 Supplier MilliporeSigma™ Supplier No. 5098800001
Add to Cart
Add to Cart

A cell-permeable inhibitor of isocitrate dehydrogenase 2 R140Q mutant-containing dimers. Has minimal effect on wild type dimer-catalyzed alpha-KG production.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins (IDH2) R140Q mutants
CAS 1432660-47-3
Molecular Formula C21H18F3N3O3S2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.