Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IgG2a from Mouse Myeloma

Catalog No. 4011231MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-112-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-112-31MG Supplier MilliporeSigma™ Supplier No. 4011231MG

missing translation for 'validMainframeMsg'

Derived from mouse myeloma.

  • Preservative ≤0.1% sodium azide

Recommended Storage

−20°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Formulation In 10mM PBS, pH 7.4.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.