Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IM-412

Catalog No. 40125110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-125-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-125-110MG Supplier MilliporeSigma™ Supplier No. 40125110MG
Add to Cart
Add to Cart

IM-412 primarily used in Inhibition.

A cell permeable imidazoxanthine shown to decrease TGF-β stimulated reporter activity in a dose-dependent manner in HEK293 reporter cells, with an IC50 of 4.1μM without cell cytotoxicity. Inhibits TGF-β receptor I (TβRI) and TGF-β receptor II (TβRII) expression and its downstream Smad2 and Smad3 phosphorylation as well as JNK and ERK activity. In addition, it elicits TGF-β-induced fibroblast differentiation with attenuated expression of several fibrotic markers such as α-SMA and fibronectin, and suppresses collagen accumulation in CCD-18Lu human normal lung fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by HPLC
Molecular Formula C16H14ClN5O2
Formula Weight 343.77g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.