Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Indirubin-3'-monoxime-5-sulphonic Acid

Catalog No. 4020881MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-208-81MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-208-81MG Supplier MilliporeSigma™ Supplier No. 4020881MG
Add to Cart
Add to Cart

A potent, reversible, and selective inhibitor of Cdk1 (IC50 = 5 nM) and Cdk5 (IC50 = 7 nM) and glycogen synthase kinase-3β (GSK-3β; IC50 = 80 nM).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Red
Description 97% by HPLC
Target Proteins CDK1
CAS 331467-05-1
Molecular Formula C16H11N3O5S
Formula Weight 357.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.