Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Innolink™ Biotin 354S

Catalog No. 20311910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-311-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-311-910MG Supplier MilliporeSigma™ Supplier No. 20311910MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Modified biotin conjugate linked to bis-aryl hydrazone chromophore, aromatic succinimidyl ester.

EMD Millipore Calbiochem™ Innolink™ Biotin 354S links by long-chain PEG4 linker.

  • Chromophore allows for direct spectrophotometric quantitation of total incorporated biotin
  • PEG4 linker preserves biotin/avidin affinity as well as increases solubility
  • Aromatic succinimidyl ester allows for higher efficiency modification of amines in aqueous buffers
  • Molecular Formula: C38H50N8O10S
  • Packaging: Glass Bottle

Recommended Storage

2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.