Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ AEBSF, Hydrochloride

Catalog No. 5084360001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-843-60001 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-843-60001 Supplier MilliporeSigma™ Supplier No. 5084360001
Add to Cart
Add to Cart

A 50 mM (50 mg/4.17 ml) sterile-filtered solution of AEBSF, Hydrochloride in H2O.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (50mg/4.17mL) sterile-filtered solution of AEBSF, Hydrochloride in H2O.
Description 97% by HPLC
Target Proteins serine proteases
Molecular Formula C8H10FNO2S·HCl
Formula Weight 239.7g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.