Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ AG 1478

Catalog No. 6585481MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-854-81MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-854-81MG Supplier MilliporeSigma™ Supplier No. 6585481MG
Add to Cart
Add to Cart

Highly potent and specific inhibitor of the epidermal growth factor (EGF) receptor tyrosine kinase (IC50 = 3nM)

Highly potent and specific inhibitor of the epidermal growth factor (EGF) receptor tyrosine kinase (IC50 = 3nM). Much higher concentrations are required for inhibition of the kinase activity of the closely related HER2 ( neu/erb -B2) receptor (IC50 > 100μM), the platelet-derived growth factor (PDGF) receptor (IC50 > 100μM), and p210Bcr-Abl (IC50 >50μM). Its mechanism of inhibition likely mimics that of the related compound, 4-(3-chloroanilino)quinazoline (CAQ), a competitive inhibitor with respect to ATP.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear to Faint Tan
Formulation A 10 mM (1mg/317μl) solution of AG 1478 in DMSO.
Description 98% by HPLC
Target Proteins Epidermal growth factor receptor kinase
CAS 175178-82-2
Molecular Formula C16H14ClN3O2
Formula Weight 315.8g/mol
Content And Storage Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.