Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set V, 50X

Catalog No. 5246291ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-462-91ML 1 mL
52-462-91SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-462-91ML Supplier MilliporeSigma™ Supplier No. 5246291ML
Add to Cart
Add to Cart

A 50X cocktail of four serine/threonine and tyrosine phosphatase inhibitors.

A cocktail of four phosphatase inhibitors for the inhibition of both serine/threonine and protein tyrosine phosphatases. Available as 1mL or as a set of five 1mL vials. Each vial contains 1mL of aqueous solution with the following components: 250mm Sodium Fluoride, 50mm β-Glycerophosphate (Cat. No. 35675), 50mm Sodium Pyrophosphate Decahydrate, 50mm Sodium Orthovanadate.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In H2O.
Content And Storage Protect from light
Form Liquid
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.