Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 1-Oleoyl-2-acetyl-sn-glycerol

Catalog No. 4954141SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
25 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-541-41SET 1 Set
49-541-425MG 25 mg
49-541-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 49-541-41SET Supplier MilliporeSigma™ Supplier No. 4954141SET
Add to Cart
Add to Cart

Cell-permeable diacylglycerol analog that activates Ca2+-dependent protein kinase C.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins Ca2+-dependent protein kinase C
CAS 86390-77-4
Molecular Formula C23H42O5
Formula Weight 398.6g/mol
Solubility DMSO, EtOH
Form Oil
Quantity 1 Set
Purity 95% by TLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.