Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2

Catalog No. 1240171MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-401-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-401-71MG Supplier MilliporeSigma™ Supplier No. 1240171MG
Add to Cart
Add to Cart

The InSolution™ Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 controls the biological activity of Akt.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (1mg/181μl) solution of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 in DMSO.
Description 95% by HPLC
Target Proteins Akt1, Akt2, Akt3
Molecular Formula C34H29N7O
Formula Weight 551.6g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.