Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set III, Lyophilized

Catalog No. 5246311ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-463-11ML 1 mL
52-463-11SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-463-11ML Supplier MilliporeSigma™ Supplier No. 5246311ML
Add to Cart
Add to Cart

The Phosphatase Inhibitor Cocktail Set III, Lyophilized controls the activity of Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.