Learn More
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | White |
| Product Type | Inhibitor |
| Target Proteins | γ-secretase and Notch processing Ab40 in CHO cells overexpressing wild type bAPP |
| CAS | 209986-17-4 |
| Molecular Formula | C27H24F2N4O3 |
| Formula Weight | 490.5g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 500 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.