Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Genistein, Soybean

Catalog No. 34583450MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-583-450MG 50 mg
34-583-420MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-583-450MG Supplier MilliporeSigma™ Supplier No. 34583450MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable, reversible, substrate competitive inhibitor of protein tyrosine kinases, including autophosphorylation of epidermal growth factor receptor kinase (IC50 = 2.6 μM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins EGFR kinase
CAS 446-72-0
Molecular Formula C15H10O5
Formula Weight 270.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.