Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ ATM Kinase Inhibitor

Catalog No. 1185022MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-850-22MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-850-22MG Supplier MilliporeSigma™ Supplier No. 1185022MG
Add to Cart
Add to Cart

The InSolution™ ATM Kinase Inhibitor controls the biological activity of ATM Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (2mg/506μl) solution of ATM Kinase Inhibitor in DMSO.
Description 95% by HPLC
Molecular Formula C21H17NO3S2
Formula Weight 395.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.