Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ BAY 11-7082

Catalog No. 19687110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-687-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-687-110MG Supplier MilliporeSigma™ Supplier No. 19687110MG
Add to Cart
Add to Cart

Target: TNF-a-inducible phosphorylation of IkBa

Supplied as a 100mm (10mg/483μl) solution of BAY 11-7082 (Cat. No. 196870) in DMSO.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 100 mM (10mg/483μl) solution of BAY 11-7082 in DMSO.
Description 95% by HPLC
Target Proteins TNF-α-inducible phosphorylation of IκBα
Molecular Formula C10H9NO2S
Formula Weight 207.2g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.