Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Bcl-2 Inhibitor VI, ABT-737

Catalog No. 1973345MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-733-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-733-45MG Supplier MilliporeSigma™ Supplier No. 1973345MG
Add to Cart
Add to Cart

A 25 mM (5 mg/246 μL) solution of Bcl-2 Inhibitor VI, ABT-737 in DMSO.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25 mM (5mg/246μL) solution of Bcl-2 Inhibitor VI, ABT-737 in DMSO.
Description 97% by HPLC
Target Proteins Bcl-2 family members
Molecular Formula C42H45ClN6O5S2
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.