Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™Bestatin, Hydrochloride

Catalog No. 20048810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-048-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-048-810MG Supplier MilliporeSigma™ Supplier No. 20048810MG
Add to Cart
Add to Cart

A metalloprotease inhibitor. Reversibly binds to cell surfaces and competitively inhibits surface aminopeptidases, notably aminopeptidase B and leucine aminopeptidase.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 50 mM (10mg/580μL) solution of Bestatatin, HCl in H2O.
Description 98% by HPLC
Target Proteins Metalloproteases
Formula Weight 344.8g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.