Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Casein Kinase II Inhibitor III, TBCA

Catalog No. 2188615MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-886-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-886-15MG Supplier MilliporeSigma™ Supplier No. 2188615MG
Add to Cart
Add to Cart

The InSolution™ Casein Kinase II Inhibitor III, TBCA, also referenced under CAS 934358-00-6, controls the biological activity of Casein Kinase II. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25 mM (5mg/431μL) solution of Casein Kinase II Inhibitor III, TBCA in DMSO.
Description 90% by HPLC
Target Proteins CKII
CAS 934358-00-6
Formula Weight 463.8g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.