Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Caspase-8 Inhibitor II

Catalog No. 218840250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-884-0250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-884-0250UG Supplier MilliporeSigma™ Supplier No. 218840250UG
Add to Cart
Add to Cart

The InSolution™ Caspase-8 Inhibitor II controls the biological activity of Caspase-8. This small molecule/inhibitor is primarily used for Cancer applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Z-Ile-Glu(OMe)-Thr-Asp(OMe)-CH2F*
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 5 mM (250μg/76μl) solution of Z-IETD-FMK in DMSO.
Description single spot by TLC
Target Proteins caspase-8
Molecular Formula C30H43FN4O11
Formula Weight 654.7g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 250 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.