Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Caspase Inhibitor I

Catalog No. 6276091MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-760-91MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 62-760-91MG Supplier MilliporeSigma™ Supplier No. 6276091MG
Add to Cart
Add to Cart

The InSolution™ Caspase Inhibitor I controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (1mg in 214μl) solution of Caspase Inhibitor I in DMSO.
Description Single spot by TLC
Target Proteins Fas-mediated apoptosis in Jurkat cells and staurosporine-induced cell death in corneal epithelial cells
Formula Weight 467.5g/mol
Content And Storage Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.