Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Cdk1 Inhibitor IV, RO-3306

Catalog No. 2177212MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-772-12MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-772-12MG Supplier MilliporeSigma™ Supplier No. 2177212MG
Add to Cart
Add to Cart

The InSolution™ Cdk1 Inhibitor IV, RO-3306 controls the biological activity of Cdk1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (2mg/569μL) solution of Cdk1 Inhibitor IV, RO-3306 in DMSO.
Description 95% by HPLC
Target Proteins Cdk1/B1 and Cdk1/A
Formula Weight 351.5g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.