Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ CFTR Inhibitor II, GlyH-101

Catalog No. 2196755MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-967-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-967-55MG Supplier MilliporeSigma™ Supplier No. 2196755MG
Add to Cart
Add to Cart

A cell-permeable glycinyl hydrazone compound that acts as a potent, selective and reversible open-channel blocker of CFTR with intermediate speed (<1 min; 95% inhibition at 50μM; Ki

A cell-permeable glycinyl hydrazone compound that acts as a potent, selective and reversible open-channel blocker of CFTR with intermediate speed (<1 min; 95% inhibition at 50μM; Ki = 4.3μM in CFTR-expressing FRT cells for apical membrane Cl- current) and exhibits desirable aqueous solubility. Shown to produce inwardly rectifying CFTR Cl- currents with reduced mean channel open time and suggested to directly interact with the channel pore at the extracellular side of the membrane. Displays minimal effects on P-glycoprotein and non-CFTR-mediated Cl- currents, and is effective in nasal and intestinal epithelia in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25 mM (5mg/406μL) solution of CFTR Inhibitor II, GlyH-101 in DMSO.
Description 97% by HPLC
Target Proteins CFTR
Molecular Formula C19H15Br2N3O3
Formula Weight 493.2g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.