Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Cyclopamine, V. californicum

Catalog No. 2398065MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-980-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-980-65MG Supplier MilliporeSigma™ Supplier No. 2398065MG
Add to Cart
Add to Cart

The InSolution™ Cyclopamine, V. californicum controls the biological activity of Sonic Hedgehog signaling pathway. This small molecule/inhibitor is primarily used for Cancer applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 10 mM (5mg/1.22mL) solution of Cyclopamine, V. californicum in EtOH.
Description 97% by NMR
Molecular Formula C27H41NO2
Formula Weight 411.6g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.