Learn More
MilliporeSigma™ Calbiochem™ InSolution™ eIF4E/eIF4G Interaction Inhibitor, 4EGI-1
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | A 25 mM (5mg/443μL) solution of eIF4E/eIF4G Interaction Inhibitor, 4EGI-1 |
| Description | 95% by HPLC (sum of two isomers) |
| Product Type | Inhibitor |
| Target Proteins | eIF4E |
| CAS | 315706-13-9 |
| Molecular Formula | C18H12Cl2N4O4S |
| Formula Weight | 451.3g/mol |
| Content And Storage | Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light |
| Form | Liquid |
| Quantity | 5 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.