Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Epoxomicin, Synthetic

Catalog No. 32480150UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-480-150UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-480-150UG Supplier MilliporeSigma™ Supplier No. 32480150UG
Add to Cart
Add to Cart

InSolution™ Epoxomicin, Synthetic, is a 1 mM solution of Epoxomicin, Synthetic (Cat. No. 324800) in DMSO.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 1 mM (50μg/90μl) solution of Epoxomicin, Synthetic in DMSO.
Description 95% by HPLC
Target Proteins CT-L,T-L,PGPH activity of proteasome
Molecular Formula C28H50N4O7
Formula Weight 554.7g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.