Learn More
Description
- Packaged Under Inert Gas
- Deactivate with 5% Bleach
Specifications
Specifications
| Formulation | A 10 mM (10mg/2.13mL) solution of Fluvastatin, Sodium Salt in H2O. |
| Description | 98% by HPLC |
| Product Type | Inhibitor |
| Target Proteins | HMG-CoA reductase |
| CAS | 93957-55-2 |
| Molecular Formula | C24H25FNNaO4 |
| Formula Weight | 433.5g/mol |
| Content And Storage | Avoid Freeze-Thaw Cycles, Protect from Light |
| Form | Liquid |
| Quantity | 10 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.