Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Foxo1 Inhibitor, AS1842856

Catalog No. 5060810001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-608-10001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-608-10001 Supplier MilliporeSigma™ Supplier No. 5060810001
Add to Cart
Add to Cart

A cell-permeable oxodihydroquinoline

A cell-permeable oxodihydroquinoline that preferentially inhibits the transcription activity of Forkhead box O family member Foxo1 over that of the functionally related Foxo3a and Foxo4 (70%, 20%, and 3% inhibition, respectively, in HepG2-based reporter assays; [AS184256] = 100nM) in a dose-dependent manner (IC50 = 33nM) via direct binding of the non-Ser256-phosphorylated/active Foxo1, but not the Ser256-phosphorylated/inactive form of Foxo1. Shown to inhibit gluconeogenesis in rat heptoma Fao cultures (IC50 = 37 and 130nM against PEPCK and G6Pase mRNA level; IC50 = 43nM against glucose production) in vitro and in murine liver of both normoglycemic ICR mice (60% and 45% inhibition of hepatic G6Pase and PEPCK mRNA level, respectively; three 100mg/kg p.o. dosages at 0, 12, 24 h) and diabetic db/db mice (86%, 70%, 56%, 48%, 35%, and 31% inhibition of hepatic G6Pase, ABCG8, ABCG5, PEPCK, IL-1β, and apoCIII mRNA level, respectively; three 100mg/kg p.o. dosages at 0, 12, 24 h) during a 26-hour fasting period in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (5mg/288μL) sterile-filtered solution of Foxo1 Inhibitor, AS1842856 in DMSO.
Description 98% by HPLC
Target Proteins Foxo1
Molecular Formula C18H22FN3O3
Formula Weight 347.4g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.