Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Gö 6976

Catalog No. 3652531ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-525-31ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-525-31ML Supplier MilliporeSigma™ Supplier No. 3652531ML
Add to Cart
Add to Cart

Inhibitor of protein kinase C (PKC; IC50 = 7

Inhibitor of protein kinase C (PKC; IC50 = 7.9nM for rat brain). Selectively inhibits Ca2+-dependent PKCα-isozyme (IC50 = 2.3nM) and PKCβI (IC50 = 6.2nM). Does not affect the kinase activity of the Ca2+-independent PKC δ-, ε-, and ζ-isoenzymes even at micromolar levels. A potent antagonist of HIV-1 induction.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 500μg/ml solution of Gö 6976 in anhydrous DMSO.
Description 95% by HPLC
Target Proteins PKC
CAS 136194-77-9
Molecular Formula C24H18N4O
Formula Weight 378.4g/mol
Content And Storage Protect from light
Form Liquid
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.