Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Glutaminase Inhibitor, Compound 968

Catalog No. 5060730001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-607-30001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-607-30001 Supplier MilliporeSigma™ Supplier No. 5060730001
Add to Cart
Add to Cart

A cell-permeable benzophenanthridinone compound that acts as an allosteric and reversible inhibitor of mitochondrial glutaminase activity and represses the growth and invasive activity in glutaminase upregulated fibroblasts and in tumor cells (IC50 ≤ 10μM in NIH3T3 cells stably expressing Dbl, Cdc42-F28L, Rac-F28L or RhoC-F30L mutants and in SKBR3 and MDA-MB231 cancer cel

A cell-permeable benzophenanthridinone compound that acts as an allosteric and reversible inhibitor of mitochondrial glutaminase activity and represses the growth and invasive activity in glutaminase upregulated fibroblasts and in tumor cells (IC50 ≤ 10μM in NIH3T3 cells stably expressing Dbl, Cdc42-F28L, Rac-F28L or RhoC-F30L mutants and in SKBR3 and MDA-MB231 cancer cells). Shown to effectively shrink tumor size in P-493 B xenografted mouse model (200μg per inj. i.p., 12 days).
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (5mg/1.05mL) sterile-filtered solution of Glutaminase Inhibitor, Compound 968 in DMSO.
Description 99% by HPLC
Target Proteins mitochondrial glutaminase
Molecular Formula C27H27BrN2O
Formula Weight 475.4g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.