Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ GM 6001

Catalog No. 3642061MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-420-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-420-61MG Supplier MilliporeSigma™ Supplier No. 3642061MG
Add to Cart
Add to Cart

InSolution™ GM 6001, CAS 142880-36-2, is a 10mm (1mg/257μl) solution of GM6001 in DMSO. A potent, cell-permeable, broad-spectrum inhibitor of MMPs.

InSolution™ GM 6001, CAS 142880-36-2, is a 10mm (1mg/257μl) solution of GM6001 in DMSO. A potent, cell-permeable, broad-spectrum inhibitor of MMPs.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (1mg/257μl) solution of GM6001 in DMSO.
Description 95% by HPLC
Target Proteins MMP-1
Molecular Formula C20H28N4O4
Formula Weight 388.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.