Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ H-89, Dihydrochloride

Catalog No. 3719621MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-196-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-196-21MG Supplier MilliporeSigma™ Supplier No. 3719621MG
Add to Cart
Add to Cart

InSolution™ H-89, Dihydrochloride, CAS 127243-85-0, is a 10mm solution of H-89, 2HCl in DMSO. A cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinase A (Ki = 48nM).

InSolution™ H-89, Dihydrochloride, CAS 127243-85-0, is a 10mm solution of H-89, 2HCl in DMSO. A cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinase A (Ki = 48nM).
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (1mg/193μl) solution of H-89, 2HCl in DMSO.
Description 99% by HPLC
Target Proteins PKA
Molecular Formula C20H20BrN3O2S
Formula Weight 519.3g/mol
Content And Storage Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.