Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ I-BET

Catalog No. 5060710001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-607-10001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-607-10001 Supplier MilliporeSigma™ Supplier No. 5060710001
Add to Cart
Add to Cart

A 50 mM sterile-filtered solution of I-BET in DMSO.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (5mg/236μL) sterile-filtered solution of I-BET in DMSO.
Description 98% by Chiral HPLC
Target Proteins tandem bromodomains of BET
CAS 1260907-17-2
Molecular Formula C22H22ClN5O2
Formula Weight 423.9g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.