Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ IKK-2 Inhibitor IV

Catalog No. 401484500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-148-4500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-148-4500UG Supplier MilliporeSigma™ Supplier No. 401484500UG
Add to Cart
Add to Cart

The InSolution™ IKK-2 Inhibitor IV controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Pale Yellow
Formulation A 10 mM (500μg/179μl) solution of IKK-2 Inhibitor IV in DMSO.
Description 95% by HPLC
Target Proteins IKK-2
Molecular Formula C12H10FN3O2S
Formula Weight 279.3g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.