Learn More
Description
Specifications
Specifications
| Formulation | A 10 mM (500μg/103μL) sterile-filtered solution of IKK Inhibitor VII in DMSO. |
| Description | 97% by HPLC |
| Product Type | Inhibitor |
| Target Proteins | IKK-2, IKK complex, and IKK-1 |
| Molecular Formula | C28H29N5OS |
| Formula Weight | 483.6g/mol |
| Content And Storage | Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light |
| Form | Liquid |
| Quantity | 500 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.