Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ JAK Inhibitor I

Catalog No. 420097500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-009-7500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-009-7500UG Supplier MilliporeSigma™ Supplier No. 420097500UG
Add to Cart
Add to Cart

The InSolution™ JAK Inhibitor I, also referenced under CAS 457081-03-7, controls the biological activity of JAK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (500μg/162μl) solution of JAK Inhibitor I in DMSO.
Description 98% by HPLC
Target Proteins murine JAK1
CAS 457081-03-7
Molecular Formula C18H16FN3O
Formula Weight 309.3g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.