Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ JNK Inhibitor II

Catalog No. 4201285MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-012-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-012-85MG Supplier MilliporeSigma™ Supplier No. 4201285MG
Add to Cart
Add to Cart

The InSolution™ JNK Inhibitor II, also referenced under CAS 129-56-6, controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish Brown
Formulation A 50 mM (5mg/454μl) solution of JNK Inhibitor II in DMSO.
Description 98% by HPLC
Target Proteins JNK 1, JNK 2, JNK 3
Molecular Formula C14H8N2O
Formula Weight 220.2g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.