Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ KN-92

Catalog No. 5083140001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-831-40001 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-831-40001 Supplier MilliporeSigma™ Supplier No. 5083140001
Add to Cart
Add to Cart

Negative Control

Useful as a negative control for KN-93 (Cat. No. 422708), a Ca2+/CaM kinase II inhibitor.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 5 mM (500μg/180μL) sterile-filtered solution of KN-92 in DMSO.
Description 95% by HPLC
Molecular Formula C24H25ClN2O3S·H3PO4
Formula Weight 555g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.