Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Latrunculin A, Latrunculia magnifica

Catalog No. 42802650UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-802-650UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-802-650UG Supplier MilliporeSigma™ Supplier No. 42802650UG
Add to Cart
Add to Cart

A cell-permeable marine toxin from the Red Sea sponge Latrunculia magnifica that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (Kd = 200nM)

A cell-permeable marine toxin from the Red Sea sponge Latrunculia magnifica that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (Kd = 200nM). Also a potent inhibitor of microfilament-mediated processes in sperm, eggs, and embryos.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 1 mM (50μg/119μl) solution of Latrunculin A, Latrunculia magnifica in DMSO.
Description 85% by HPLC
Target Proteins Inhibitor of microfilament-mediated processes in sperm, eggs, and embryos
Molecular Formula C22H31NO5S
Formula Weight 421.6g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.