Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Leupeptin, Hemisulfate, Microbial

Catalog No. 5092810001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-928-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-928-10001 Supplier MilliporeSigma™ Supplier No. 5092810001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 50 mM (10 mg/421 μl) sterile-filtered solution of Leupeptin, Hemisulfate, Microbial in H2O.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Leu-Leu-arginal, hemisulfate
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (10mg/421μl) sterile-filtered solution of Leupeptin, Hemisulfate, Microbial in H2O.
Description 90% by HPLC
Target Proteins trypsin-like proteases and cysteine proteases
CAS 103476-89-7
Molecular Formula C20H38N6O4·0.5(H2SO4)
Formula Weight 475.6g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.