Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Lovastatin, Sodium Salt

Catalog No. 4381875MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-818-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-818-75MG Supplier MilliporeSigma™ Supplier No. 4381875MG
Add to Cart
Add to Cart

Carboxylate form of Lovastatin that is active in whole cells and cell-free assays.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (5mg/1.13mL) solution of Lovastatin, Sodium Salt in H2O.
Description 95% by HPLC
Target Proteins HMG-CoA reductase
Molecular Formula C24H37O6·Na
Formula Weight 444.5g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.