Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ MG-132 in EtOH, ≥95% by HPLC

Catalog No. 47478810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-478-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-478-810MG Supplier MilliporeSigma™ Supplier No. 47478810MG
Add to Cart
Add to Cart

Potent, reversible, and cell-permeable proteasome inhibitor. Reduces the degradation of ubiquitin-conjugated proteins in mammalian cells and permeable stains of yeast by the 26S complex.

  • Packaged Under Inert Gas
  • Peptide Sequence: Z-Leu-Leu-Leu-CHO
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 20 mM (10mg/1.05mL) solution ofmg-132, 95% by HPLC in EtOH.
Description 95% by HPLC
Molecular Formula C26H41N3O5
Formula Weight 475.6g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.