Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Necrostatin-1

Catalog No. 5052240001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-522-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-522-40001 Supplier MilliporeSigma™ Supplier No. 5052240001
Add to Cart
Add to Cart

A cell-permeable, potent, and selective blocker of necroptosis (EC50 = 494nM in FADD-deficient Jurkat cells treated with TNF-α), a nonapoptotic necrotic cell death pathway mediated by death-domain receptors (DRs) that offers neuroprotection in a murine model of ischemic brain inj

A cell-permeable, potent, and selective blocker of necroptosis (EC50 = 494nM in FADD-deficient Jurkat cells treated with TNF-α), a nonapoptotic necrotic cell death pathway mediated by death-domain receptors (DRs) that offers neuroprotection in a murine model of ischemic brain injury. Exhibits no effect on DR-induced apoptosis. Also acts as a selective and ATP-competitive inhibitor of RIP1 kinase with negligible effect on RIP2 kinase activity. Nec-1 target appears to be a critical common necroptotic step upstream of execution events and downstream of DRs. Inactive control, Nec-1i, is also available ( Cat. No. 480066).
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25 mM (10mg/1.54mL) sterile-filtered solution of Necrostatin-1 in DMSO.
Description 99% by HPLC
Molecular Formula C13H13N3OS
Formula Weight 259.3g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.