Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Nutlin-3, Racemic

Catalog No. 4441511MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-415-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-415-11MG Supplier MilliporeSigma™ Supplier No. 4441511MG
Add to Cart
Add to Cart

A cell-permeable, potent, and selective MDM2 antagonist (IC50 = 90nM for Nutlin-3a and 13

A cell-permeable, potent, and selective MDM2 antagonist (IC50 = 90nM for Nutlin-3a and 13.6μM for Nutlin-3b) that displays antitumor properties. Activates the p53 pathway by binding MDM2 in the p53-binding pocket and inhibiting the MDM2-p53 interaction. Shown to induce apoptosis in several cancer cell lines with wild-type p53 and in xenografted tumor mice.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (1mg/172μl) solution of MDM2 Antagonist, Nutlin-3, Racemic in DMSO
Description 98% by HPLC
Molecular Formula C30H30Cl2N4O4
Formula Weight 581.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.