Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ OM99-2

Catalog No. 496000250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-600-0250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-600-0250UG Supplier MilliporeSigma™ Supplier No. 496000250UG
Add to Cart
Add to Cart

An eight residue peptidomimetic, tight binding transition-state analog inhibitor of human brain memapsin 2.

An eight residue peptidomimetic, tight binding transition-state analog inhibitor of human brain memapsin 2 (Ki = 1.6 nM, recombinant memapsin 2; Ki = 9.58 nM, recombinant pro-memapsin 2). Also inhibits cathepsin D (Ki = 48nM). This aspartyl protease inhibitor is designed from the template of the β-secretase site of Swedish β-amyloid precursor protein (APP) with Asp to Ala replacement; also includes a nonhydrolyzable hydroxyethylene isostere between Leu and Ala.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 1 mM (250μg in 280μl) solution in DMSO. Supplied as a trifluoroacetate salt.
Description 90% by HPLC
Target Proteins human brain memapsin 2
Molecular Formula C41H64N8O14
Formula Weight 893g/mol
Content And Storage Protect from Light, Protect from Moisture
Form Liquid
Quantity 250 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.