Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Paclitaxel, Taxus sp.

Catalog No. 5082270001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-822-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-822-70001 Supplier MilliporeSigma™ Supplier No. 5082270001
Add to Cart
Add to Cart

Antitumor and anti-leukemic agent isolated from the bark of the yew tree

Antitumor and anti-leukemic agent isolated from the bark of the yew tree. Promotes assembly of microtubules and inhibits tubulin disassembly. An effective agent in blocking HeLa and B16 cells at the G2/M stage. Inhibits DNA synthesis and stimulates the release of tumor necrosis factor-α. Induces apoptosis in murine mammary carcinoma MCA-4 and ovarian carcinoma OCA-1 tumors.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25 mM (10mg/468μL) sterile-filtered solution of Paclitaxel, Taxus sp. in DMSO.
Description 97% by HPLC
Target Proteins microtubules
Molecular Formula C47H51NO14
Formula Weight 853.9g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.