Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Pepstatin A, Synthetic

Catalog No. 5084370001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-843-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-843-70001 Supplier MilliporeSigma™ Supplier No. 5084370001
Add to Cart
Add to Cart

A 25 μM (10 mg/583 μL) sterile-filtered solution of Pepstatin A, Synthetic in DMSO.

  • Packaged Under Inert Gas
  • Peptide Sequence: Iva-Val-Val-Sta-Ala-Sta
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25μM (10mg/583μL) sterile-filtered solution of Pepstatin A, Synthetic in DMSO.
Description 98% by HPLC
Target Proteins aspartic proteases
Molecular Formula C34H63N5O9
Formula Weight 685.9g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.