Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ PERK Inhibitor I, GSK2606414

Catalog No. 5083400001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-834-00001 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-834-00001 Supplier MilliporeSigma™ Supplier No. 5083400001
Add to Cart
Add to Cart

A cell-permeable pyrrolopyrimidinamine compound that acts as a highly potent inhibitor against EIF2AK3/PERK-catalyzed EIF2α Ser51 phosphorylation in kinase assays (IC50 = 0

A cell-permeable pyrrolopyrimidinamine compound that acts as a highly potent inhibitor against EIF2AK3/PERK-catalyzed EIF2α Ser51 phosphorylation in kinase assays (IC50 = 0.4nM; 30 min preincubation; [ATP] = 5μM) as well as ER stress-induced PERK autophosphorylation following Thapsigargin (Cat. No. 586005) addition in A549 cultures (by 100% with ≤30nM inhibitor; 60 min preincubation) by targeting PERK in its inactive conformation at the ATP-binding region. Displays ≥385-fold selectivity over c-Kit, Aurora B, BRK, HRI/EIF2AK1, MLK2/MAP3K10, c-MER, DDR2, PKR/EIF2AK2, and MLCK2/MYLK2 (IC50 = 154, 407, 412, 420, 452, 474, 524, 696, and 701 nM, respectively) and exhibits much reduced or little activity against more than 280 other kinases (IC50 >1μM). Reported to be orally available in dog, mouse, and rat with good pharmacokinetics and retard the growth of established PxBC-3 tumor mass in mice in vivo (by 20% and 59% at the end of a 21 day treatment period with b.i.d. oral dosage of 50 and 150mg/kg, respectively). The preincubation time-dependent inhibition, extremely slow dissociation rate, as well as the observed PERK selectivity, are all consistent with the inhibitor targeting PERK in its inactive DFG conformation.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25 mM (2mg/177μL) sterile-filtered solution of PERK Inhibitor I, GSK2606414 in DMSO.
Description 98% by HPLC
Target Proteins PERK
Molecular Formula C24H20F3N5O
Formula Weight 451.4g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.